Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPEPLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122838.1 | NP_001116310.1 | 2 | 468 | ||
NM_198990.4 | NP_945341.3 | 2 | 468 | |||
XM_005250214.4 | XP_005250271.2 | 2 | 200 | |||
XM_005250215.2 | XP_005250272.1 | 2 | 333 | |||
XM_005250218.2 | XP_005250275.1 | 2 | 300 | |||
XM_011515933.2 | XP_011514235.1 | 2 | 200 | |||
XM_017011854.1 | XP_016867343.1 | 2 | 591 | |||
XM_017011855.1 | XP_016867344.1 | 2 | 333 | |||
XM_017011856.1 | XP_016867345.1 | 2 | 438 | |||
XM_017011857.1 | XP_016867346.1 | 2 | 438 | |||
GenBank mRNA | AB112352.1 | - | 1 | 140 | ||
AK000801.1 | - | 2 | 357 | |||
AK304644.1 | - | 2 | 438 | |||
AY357337.1 | - | 2 | 234 | |||
BC071604.1 | - | 2 | 430 | 4375696 | ||
CR936639.1 | - | 2 | 419 |