Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145128.2 | NP_001138600.2 | 39 | 5460 | ||
XM_006715376.3 | XP_006715439.2 | 40 | 6325 | |||
XM_011535550.2 | XP_011533852.1 | 40 | 6340 | |||
XM_011535552.2 | XP_011533854.1 | 39 | 5516 | |||
XM_011535553.2 | XP_011533855.1 | 39 | 6123 | |||
XM_011535554.2 | XP_011533856.1 | 38 | 6019 | |||
XM_011535558.1 | XP_011533860.1 | 21 | 3361 | |||
XM_017010385.1 | XP_016865874.1 | 39 | 5508 | |||
XM_017010387.1 | XP_016865876.1 | 34 | 5971 | |||
XM_017010389.1 | XP_016865878.1 | 25 | 4223 | |||
GenBank mRNA | AK098657.1 | - | 5 | 698 | ||
BC101714.1 | - | 5 | 626 | 8069220 | ||
BC111948.1 | - | 5 | 626 | 8327416 | ||
BC143533.1 | - | 5 | 584 | 9052042 |