Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HS3ST5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153612.3 | NP_705840.2 | 2 | 689 | ||
XM_006715379.2 | XP_006715442.1 | 5 | 1177 | |||
XM_011535588.2 | XP_011533890.1 | 3 | 1893 | |||
XM_017010470.1 | XP_016865959.1 | 5 | 1751 | |||
XM_017010471.1 | XP_016865960.1 | 5 | 2710 | |||
XM_017010472.1 | XP_016865961.1 | 4 | 2347 | |||
XM_017010473.1 | XP_016865962.1 | 5 | 1747 | |||
XM_017010474.1 | XP_016865963.1 | 5 | 1185 | |||
GenBank mRNA | AF503292.1 | - | 2 | 657 | ||
AK091074.1 | - | 1 | 206 | |||
AK126930.1 | - | 2 | 1349 | |||
AK289907.1 | - | 5 | 1176 | |||
BC093911.1 | - | 2 | 690 | 7939756 | ||
BC093913.1 | - | 2 | 690 | 7939758 |