Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134450.1 | NP_001127922.1 | 4 | 773 | ||
NM_001134451.1 | NP_001127923.1 | 3 | 467 | |||
NM_152913.2 | NP_690877.1 | 4 | 773 | |||
GenBank mRNA | AK124634.1 | - | 4 | 1650 | ||
AK225783.1 | - | 4 | 716 | |||
AK294658.1 | - | 4 | 589 | |||
AK295543.1 | - | 3 | 431 | |||
AK312364.1 | - | 4 | 643 | |||
AL832523.1 | - | 3 | 507 | |||
AL834348.1 | - | 4 | 619 | |||
AL834349.1 | - | 4 | 589 | |||
AY358583.1 | - | 5 | 705 | |||
BC030793.1 | - | 4 | 742 | 5263130 | ||
BC037895.1 | - | 3 | 379 | 5275566 |