Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040661.1 | NP_001035751.1 | 9 | 1253 | ||
NM_001300847.1 | NP_001287776.1 | 9 | 1208 | |||
NM_153247.2 | NP_694979.2 | 9 | 1250 | |||
XM_005249658.4 | XP_005249715.1 | 9 | 1174 | |||
XM_006715667.3 | XP_006715730.1 | 9 | 1121 | |||
XM_006715668.2 | XP_006715731.1 | 9 | 1199 | |||
XM_011515200.2 | XP_011513502.1 | 9 | 1281 | |||
XM_011515201.2 | XP_011513503.1 | 9 | 1270 | |||
GenBank mRNA | AK075422.1 | - | 7 | 827 | ||
AK092242.1 | - | 9 | 1236 | |||
AY485959.1 | - | 9 | 1250 | |||
BC025325.1 | - | 9 | 1109 | 3354432 | ||
BC047592.1 | - | 9 | 1214 | 4816109 |