Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173565.3 | NP_775836.3 | 9 | 1204 | ||
XM_005249659.3 | XP_005249716.1 | 9 | 1706 | |||
XM_011515203.1 | XP_011513505.1 | 10 | 1286 | |||
XM_011515204.2 | XP_011513506.1 | 9 | 1474 | |||
XM_011515207.1 | XP_011513509.1 | 9 | 1166 | |||
XM_011515208.2 | XP_011513510.1 | 9 | 1706 | |||
XM_011515209.2 | XP_011513511.1 | 9 | 1706 | |||
XM_011515210.1 | XP_011513512.1 | 3 | 448 | |||
XM_011515212.2 | XP_011513514.1 | 9 | 1706 | |||
XM_017011870.1 | XP_016867359.1 | 9 | 1706 | |||
GenBank mRNA | BC034495.1 | - | 9 | 1207 | 4825868 | |
BC157864.1 | - | 7 | 954 | 9057083 | ||
BC157871.1 | - | 7 | 954 | 9057090 |
Gene Symbol: RSPH10B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099697.1 | NP_001093167.1 | 9 | 1204 | ||
XM_005249833.3 | XP_005249890.1 | 3 | 396 | |||
XM_006715766.2 | XP_006715829.1 | 10 | 1341 | |||
XM_011515491.1 | XP_011513793.1 | 9 | 1233 | |||
XM_011515493.1 | XP_011513795.1 | 9 | 1166 | |||
GenBank mRNA | AK093411.1 | - | 4 | 541 | ||
AK294481.1 | - | 4 | 502 | |||
AK314983.1 | - | 8 | 1324 | |||
BC044242.1 | - | 5 | 531 |