Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH10B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099697.1 | NP_001093167.1 | 9, 10 | 1232 | ||
GenBank mRNA | AK093411.1 | - | 4, 5 | 569 | ||
AK294481.1 | - | 4, 5 | 530 | |||
AK314983.1 | - | 8, 9 | 1352 | |||
BC044242.1 | - | 5, 6 | 559 |
Gene Symbol: RSPH10B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173565.3 | NP_775836.3 | 9, 10 | 1232 | ||
XM_005249659.3 | XP_005249716.1 | 9, 10 | 1734 | |||
XM_011515209.2 | XP_011513511.1 | 9, 10 | 1734 | |||
GenBank mRNA | BC034495.1 | - | 9, 10 | 1235 | 4825868 | |
BC157864.1 | - | 7, 8 | 982 | 9057083 | ||
BC157871.1 | - | 7, 8 | 982 | 9057090 |