Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153453.1 | NP_703154.1 | 1, 2 | 291 | ||
NM_182645.3 | NP_872586.1 | 1, 2 | 291 | |||
XM_005266883.2 | XP_005266940.1 | 1, 2 | 108 | |||
GenBank mRNA | AJ578053.1 | - | 1, 2 | 275 | ||
AK096878.1 | - | 1, 2 | 108 | |||
AY056583.1 | - | 1, 2 | 291 | |||
BC069316.1 | - | 1, 2 | 168 | |||
BC100798.2 | - | 1, 2 | 258 | |||
BC100799.1 | - | 1, 2 | 258 | |||
BC118545.1 | - | 1, 2 | 258 | |||
BC118622.1 | - | 1, 2 | 258 |