Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037497.1 | NP_001032586.1 | 2 | 221 | ||
GenBank mRNA | DQ012015.1 | - | 2 | 221 |