Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037730.1 | NP_001032819.1 | 2 | 109 | ||
GenBank mRNA | DQ012019.1 | - | 2 | 109 |