Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037804.1 | NP_001032893.1 | 2 | 140 | ||
GenBank mRNA | DQ012022.1 | - | 2 | 140 |
Gene Symbol: LOC100133267 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195257.1 | NP_001182186.1 | 2 | 140 |