Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASEH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286834.1 | NP_001273763.1 | 6 | 745 | ||
NM_001286837.1 | NP_001273766.1 | 6 | 983 | |||
NM_002936.4 | NP_002927.2 | 6 | 745 | |||
GenBank mRNA | AF039652.1 | - | 6 | 715 | ||
AF048994.1 | - | 6 | 661 | |||
AF048995.1 | - | 6 | 695 | |||
AJ224117.1 | - | 6 | 634 | |||
AK096913.1 | - | 6 | 901 | |||
BC002973.1 | - | 6 | 704 | 3537074 | ||
BT019670.1 | - | 6 | 634 | |||
CR541820.1 | - | 6 | 634 |