Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145315.4 | NP_660358.2 | 11 | 1376 | ||
NR_136553.1 | 7 | 939 | ||||
XM_005266885.3 | XP_005266942.1 | 12 | 2734 | |||
XM_011535657.2 | XP_011533959.1 | 10 | 1157 | |||
XM_017010647.1 | XP_016866136.1 | 12 | 1226 | |||
XM_017010648.1 | XP_016866137.1 | 10 | 828 | |||
XM_017010649.1 | XP_016866138.1 | 11 | 1492 | |||
GenBank mRNA | AF520418.1 | - | 11 | 1356 | ||
BC018445.1 | - | 11 | 1356 | 3871205 | ||
BI754108.1 | - | 7 | 634 |