Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LACE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323005.1 | NP_001309934.1 | 7, 8 | 1018 | ||
NM_145315.4 | NP_660358.2 | 7, 8 | 1018 | |||
XM_005266885.3 | XP_005266942.1 | 8, 9 | 2376 | |||
XM_011535657.2 | XP_011533959.1 | 6, 7 | 799 | |||
XM_017010647.1 | XP_016866136.1 | 8, 9 | 868 | |||
XM_017010648.1 | XP_016866137.1 | 6, 7 | 470 | |||
XM_017010649.1 | XP_016866138.1 | 7, 8 | 1134 | |||
GenBank mRNA | AF520418.1 | - | 7, 8 | 998 | ||
BC018445.1 | - | 7, 8 | 998 | 3871205 |