Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286256.1 | NP_001273185.1 | 2 | 216 | ||
NM_145064.2 | NP_659501.1 | 3 | 283 | |||
XM_011538126.2 | XP_011536428.1 | 3 | 371 | |||
GenBank mRNA | AK057013.1 | - | 3 | 276 | ||
BC008069.2 | - | 2 | 196 | 2961089 | ||
DA896836.1 | - | 3 | 283 |