Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098510.1 | NP_001091980.1 | 7 | 1360 | ||
NM_001243135.1 | NP_001230064.1 | 7 | 1199 | |||
NM_001243136.1 | NP_001230065.1 | 6 | 1207 | |||
NM_145065.2 | NP_659502.2 | 8 | 1432 | |||
XM_011544884.2 | XP_011543186.1 | 8 | 1435 | |||
XM_017017465.1 | XP_016872954.1 | 6 | 1184 | |||
GenBank mRNA | AF487456.1 | - | 7 | 1268 | ||
AF487457.1 | - | 6 | 1196 | |||
AK074201.1 | - | 8 | 1370 | |||
AK075253.1 | - | 4 | 872 | |||
AK094060.1 | - | 7 | 1199 | |||
AL834395.1 | - | 8 | 1379 | |||
BC025723.1 | - | 7 | 1281 | 5198999 | ||
BC127759.1 | - | 2 | 518 |