Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145659.3 | NP_663634.2 | 5 | 927 | ||
XM_011545780.2 | XP_011544082.1 | 5 | 1281 | |||
GenBank mRNA | BC062422.1 | - | 5 | 930 | 4703457 |