Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288564.1 | NP_001275493.1 | 3 | 271 | ||
NM_001288565.1 | NP_001275494.1 | 2 | 774 | |||
NM_001288566.1 | NP_001275495.1 | 3 | 372 | |||
NM_001288567.1 | NP_001275496.1 | 3 | 586 | |||
NM_001288568.1 | NP_001275497.1 | 3 | 424 | |||
NM_001288569.1 | NP_001275498.1 | 3 | 285 | |||
NM_001288570.1 | NP_001275499.1 | 3 | 298 | |||
NM_001288571.1 | NP_001275500.1 | 3 | 364 | |||
NM_016456.4 | NP_057540.1 | 3 | 289 | |||
NR_109920.1 | 3 | 289 | ||||
NR_109921.1 | 2 | 774 | ||||
XM_011509385.1 | XP_011507687.1 | 3 | 348 | |||
XM_011509386.2 | XP_011507688.1 | 3 | 323 | |||
XM_011509387.2 | XP_011507689.1 | 3 | 211 | |||
GenBank mRNA | AB013909.1 | - | 2 | 142 | ||
AF151020.1 | - | 4 | 261 | |||
AK054883.1 | - | 3 | 244 | |||
AK075335.1 | - | 3 | 199 | |||
AK091726.1 | - | 2 | 659 | |||
AK296307.1 | - | 3 | 206 | |||
AK303843.1 | - | 3 | 310 | |||
AL136600.1 | - | 3 | 222 | |||
AL516746.3 | - | 3 | 464 | |||
AY138587.1 | - | 3 | 200 | |||
AY359012.1 | - | 3 | 263 | |||
BC001106.2 | - | 3 | 213 | 3506442 | ||
BE782946.1 | - | 3 | 251 | |||
BM928361.1 | - | 3 | 211 | |||
BX337142.2 | - | 2 | 658 | |||
CR457162.1 | - | 2 | 142 | |||
CR533467.1 | - | 2 | 142 | |||
CX751563.1 | - | 2 | 656 | |||
DA141558.1 | - | 3 | 271 | |||
DA364005.1 | - | 3 | 289 | |||
DB031447.1 | - | 3 | 265 |