Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288564.1 | NP_001275493.1 | 5 | 505 | ||
NM_001288565.1 | NP_001275494.1 | 4 | 1008 | |||
NM_001288566.1 | NP_001275495.1 | 5 | 606 | |||
NM_001288567.1 | NP_001275496.1 | 5 | 820 | |||
NM_001288568.1 | NP_001275497.1 | 5 | 658 | |||
NM_001288569.1 | NP_001275498.1 | 5 | 519 | |||
NM_001288570.1 | NP_001275499.1 | 5 | 532 | |||
NM_001288571.1 | NP_001275500.1 | 5 | 598 | |||
NM_016456.4 | NP_057540.1 | 5 | 523 | |||
NR_109920.1 | 6 | 750 | ||||
NR_109921.1 | 4 | 977 | ||||
XM_011509385.1 | XP_011507687.1 | 5 | 582 | |||
XM_011509386.2 | XP_011507688.1 | 5 | 557 | |||
XM_011509387.2 | XP_011507689.1 | 5 | 445 | |||
GenBank mRNA | AF151020.1 | - | 6 | 495 | ||
AK054883.1 | - | 5 | 478 | |||
BM928361.1 | - | 6 | 672 | |||
BX337142.2 | - | 4 | 861 |