Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171941.2 | NP_001165412.1 | 5 | 546 | ||
NM_153756.2 | NP_715637.2 | 5 | 612 | |||
GenBank mRNA | AK092102.1 | - | 5 | 545 | ||
BC062297.1 | - | 5 | 535 | 6338770 | ||
DA393478.1 | - | 3 | 216 |