Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284334.1 | NP_001271263.1 | 7 | 594 | ||
NM_014509.4 | NP_055324.2 | 9 | 786 | |||
XM_017028738.1 | XP_016884227.1 | 9 | 814 | |||
XM_017028739.1 | XP_016884228.1 | 8 | 731 | |||
GenBank mRNA | AK295223.1 | - | 9 | 743 | ||
AL450314.1 | - | 9 | 729 | |||
AL589866.1 | - | 3 | 87 | |||
AL590118.1 | - | 7 | 435 | |||
AL590120.1 | - | 9 | 627 | |||
BC119650.2 | - | 9 | 670 | 40115836 | ||
BC119651.1 | - | 9 | 670 | 40115837 | ||
BC127635.1 | - | 9 | 671 | 40115838 | ||
BC127636.1 | - | 9 | 671 | 40115841 | ||
CR456455.1 | - | 3 | 93 |