Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERHL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284334.1 | NP_001271263.1 | 9 | 732 | ||
NM_014509.4 | NP_055324.2 | 11 | 924 | |||
NR_104300.1 | 10 | 1603 | ||||
NR_104301.1 | 10 | 1587 | ||||
XM_017028738.1 | XP_016884227.1 | 11 | 952 | |||
XM_017028739.1 | XP_016884228.1 | 10 | 869 | |||
GenBank mRNA | AK294584.1 | - | 10 | 1547 | ||
AL450314.1 | - | 11 | 867 | |||
AL589866.1 | - | 5 | 225 | |||
AL590118.1 | - | 9 | 573 | |||
AL590120.1 | - | 11 | 765 | |||
BC119650.2 | - | 11 | 808 | 40115836 | ||
BC119651.1 | - | 11 | 808 | 40115837 | ||
BC127635.1 | - | 11 | 809 | 40115838 | ||
BC127636.1 | - | 11 | 809 | 40115841 | ||
CR456455.1 | - | 5 | 231 |