Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RICTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285439.1 | NP_001272368.1 | 26 | 2587 | ||
NM_001285440.1 | NP_001272369.1 | 26 | 2585 | |||
NM_152756.4 | NP_689969.2 | 26 | 2587 | |||
XM_006714463.3 | XP_006714526.1 | 26 | 2600 | |||
XM_011514005.2 | XP_011512307.1 | 26 | 2600 | |||
XM_011514006.2 | XP_011512308.1 | 25 | 2404 | |||
XM_017009311.1 | XP_016864800.1 | 25 | 2520 | |||
XM_017009312.1 | XP_016864801.1 | 25 | 2520 | |||
XM_017009313.1 | XP_016864802.1 | 25 | 2541 | |||
XM_017009314.1 | XP_016864803.1 | 25 | 2510 | |||
XM_017009315.1 | XP_016864804.1 | 25 | 2510 | |||
GenBank mRNA | AB082530.1 | - | 11 | 1258 | ||
AY515854.1 | - | 26 | 2583 | |||
BC051729.1 | - | 6 | 585 | |||
BC137163.1 | - | 26 | 2580 | 9021160 | ||
BC137164.1 | - | 26 | 2580 | 9021161 | ||
BC144509.1 | - | 26 | 2580 | 9053036 | ||
CR749280.1 | - | 26 | 2566 |