Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000126.3 | NP_000117.1 | 3, 4 | 355 | ||
NM_001127716.1 | NP_001121188.1 | 2, 3 | 208 | |||
GenBank mRNA | AK292979.1 | - | 3, 4 | 355 | ||
AK300044.1 | - | 2, 3 | 143 | |||
BC015526.1 | - | 3, 4 | 299 | 3852346 | ||
BC029995.1 | - | 3, 4 | 344 | |||
BC095457.1 | - | 3, 4 | 280 | 30552923 | ||
BT009796.1 | - | 3, 4 | 274 | |||
BU176141.1 | - | 2, 3 | 171 | |||
DA838974.1 | - | 3, 4 | 355 | |||
J04058.1 | - | 3, 4 | 274 | |||
KU178046.1 | - | 3, 4 | 274 | |||
KU178047.1 | - | 4, 5 | 407 | |||
KU178048.1 | - | 3, 4 | 274 |