Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278505.1 | NP_001265434.1 | 4, 5 | 400 | ||
NM_001322249.1 | NP_001309178.1 | 5, 6 | 627 | |||
NM_174931.3 | NP_777591.3 | 6, 7 | 669 | |||
XM_017003748.1 | XP_016859237.1 | 6, 7 | 570 | |||
GenBank mRNA | AK095667.1 | - | 4, 5 | 333 | ||
AK289504.1 | - | 4, 5 | 315 | |||
AK294697.1 | - | 6, 7 | 583 | |||
BC071798.1 | - | 4, 5 | 332 | |||
BM285380.1 | - | 6, 7 | 669 |