Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281737.1 | NP_001268666.1 | 4 | 732 | ||
NM_001281738.1 | NP_001268667.1 | 3 | 429 | |||
NM_004453.3 | NP_004444.2 | 5 | 873 | |||
GenBank mRNA | AK293737.1 | - | 3 | 414 | ||
AK304838.1 | - | 4 | 494 | |||
AK314803.1 | - | 5 | 850 | |||
BC011890.1 | - | 5 | 675 | 3938660 | ||
DB221748.1 | - | 3 | 431 | |||
EU093082.1 | - | 5 | 569 | |||
S69232.1 | - | 5 | 637 |