Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286638.1 | NP_001273567.1 | 9 | 1038 | ||
NM_153251.3 | NP_694983.2 | 10 | 1064 | |||
NR_104486.1 | 11 | 1183 | ||||
NR_104487.1 | 10 | 1065 | ||||
XM_006719795.1 | XP_006719858.1 | 10 | 1004 | |||
XM_006719796.1 | XP_006719859.1 | 10 | 1004 | |||
XM_006719797.1 | XP_006719860.1 | 10 | 1004 | |||
XM_011535030.2 | XP_011533332.1 | 4 | 483 | |||
XM_017020525.1 | XP_016876014.1 | 8 | 697 | |||
XM_017020526.1 | XP_016876015.1 | 11 | 770 | |||
XM_017020527.1 | XP_016876016.1 | 10 | 980 | |||
GenBank mRNA | AK295804.1 | - | 10 | 970 | ||
AK299354.1 | - | 9 | 976 | |||
BC034944.1 | - | 12 | 1407 | |||
BC050367.1 | - | 10 | 979 | 5591379 | ||
BC067898.1 | - | 11 | 1182 | |||
BC110517.1 | - | 10 | 943 | 40032074 |