Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286638.1 | NP_001273567.1 | 2, 3 | 427 | ||
NM_153251.3 | NP_694983.2 | 3, 4 | 453 | |||
NR_104486.1 | 3, 4 | 454 | ||||
NR_104487.1 | 3, 4 | 454 | ||||
XM_006719795.1 | XP_006719858.1 | 3, 4 | 393 | |||
XM_006719796.1 | XP_006719859.1 | 3, 4 | 393 | |||
XM_006719797.1 | XP_006719860.1 | 3, 4 | 393 | |||
GenBank mRNA | AK295804.1 | - | 3, 4 | 359 | ||
AK299354.1 | - | 2, 3 | 365 | |||
BC034944.1 | - | 3, 4 | 459 | |||
BC050367.1 | - | 3, 4 | 368 | 5591379 | ||
BC067898.1 | - | 3, 4 | 453 | |||
BC110517.1 | - | 3, 4 | 332 | 40032074 | ||
BG720734.1 | - | 3, 4 | 459 |
Gene Symbol: UBIAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK074487.1 | - | 292 |