Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETFDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281737.1 | NP_001268666.1 | 11 | 1731 | ||
NM_001281738.1 | NP_001268667.1 | 10 | 1428 | |||
NM_004453.3 | NP_004444.2 | 12 | 1872 | |||
GenBank mRNA | AK293737.1 | - | 10 | 1413 | ||
AK304838.1 | - | 11 | 1493 | |||
AK314803.1 | - | 12 | 1849 | |||
BC011890.1 | - | 12 | 1674 | 3938660 | ||
BX538129.1 | - | 13 | 1544 | |||
EU093082.1 | - | 12 | 1568 | |||
EU093083.1 | - | 4 | 486 | |||
S69232.1 | - | 12 | 1636 |