Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277813.1 | NP_001264742.1 | 5 | 1854 | ||
NM_001277814.1 | NP_001264743.1 | 5 | 1608 | |||
NM_001277815.1 | NP_001264744.1 | 5 | 1739 | |||
NM_001277816.1 | NP_001264745.1 | 3 | 1622 | |||
NM_001277817.1 | NP_001264746.1 | 4 | 1547 | |||
NM_001277818.1 | NP_001264747.1 | 4 | 1468 | |||
NM_152758.5 | NP_689971.4 | 4 | 1708 | |||
XM_011517509.2 | XP_011515811.1 | 5 | 1701 | |||
XM_011517510.2 | XP_011515812.1 | 3 | 1464 | |||
GenBank mRNA | AK056219.1 | - | 4 | 1618 | ||
AK093081.1 | - | 4 | 1468 | |||
AK127574.1 | - | 4 | 1640 | |||
AK298544.1 | - | 5 | 1589 | |||
AL831816.1 | - | 4 | 1543 | |||
AL832005.1 | - | 4 | 1641 | |||
AL832369.1 | - | 2 | 1342 | |||
BC052970.1 | - | 4 | 1657 | 6044054 | ||
BX537996.2 | - | 4 | 1645 |