Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178863.4 | NP_849194.1 | 5 | 926 | ||
NR_110933.1 | 5 | 1464 | ||||
XM_011545783.2 | XP_011544085.1 | 5 | 995 | |||
XM_011545784.2 | XP_011544086.1 | 6 | 1014 | |||
XM_017023104.1 | XP_016878593.1 | 6 | 960 | |||
GenBank mRNA | AF289573.1 | - | 5 | 873 | ||
AF401315.2 | - | 5 | 838 | |||
AK097407.1 | - | 5 | 1069 | |||
AK289630.1 | - | 5 | 903 | |||
AY027918.1 | - | 5 | 850 | |||
BC036228.1 | - | 5 | 918 | 5276078 |