Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143820.1 | NP_001137292.1 | 4 | 367 | ||
NM_005238.3 | NP_005229.1 | 2 | 466 | |||
XM_011542649.2 | XP_011540951.1 | 5 | 503 | |||
XM_011542650.1 | XP_011540952.1 | 3 | 958 | |||
XM_011542651.1 | XP_011540953.1 | 4 | 398 | |||
XM_011542652.1 | XP_011540954.1 | 2 | 856 | |||
XM_017017314.1 | XP_016872803.1 | 4 | 356 | |||
XM_017017315.1 | XP_016872804.1 | 3 | 282 | |||
XM_017017316.1 | XP_016872805.1 | 3 | 16473 | |||
XM_017017317.1 | XP_016872806.1 | 4 | 402 | |||
GenBank mRNA | AK291840.1 | - | 2 | 460 | ||
AK301405.1 | - | 2 | 465 | |||
BC017314.2 | - | 2 | 420 | 3946751 | ||
BP365215.1 | - | 2 | 464 | |||
BT019452.1 | - | 2 | 150 | |||
BT019453.1 | - | 2 | 150 | |||
BX640634.1 | - | 4 | 367 | |||
CR542254.1 | - | 2 | 150 | |||
J04101.1 | - | 2 | 269 | |||
S87905.1 | - | 198 | ||||
X14798.1 | - | 2 | 428 |