Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153252.4 | NP_694984.4 | 24 | 3047 | ||
XM_005262113.3 | XP_005262170.1 | 23 | 2899 | |||
XM_017029384.1 | XP_016884873.1 | 20 | 2577 | |||
GenBank mRNA | AK095887.1 | - | 7 | 920 | ||
AY497046.1 | - | 24 | 3047 | |||
AY497047.1 | - | 21 | 2979 | |||
AY497048.1 | - | 24 | 3070 | |||
AY497049.1 | - | 23 | 3040 | |||
AY497050.1 | - | 24 | 3020 | |||
AY497051.1 | - | 21 | 2607 | |||
AY497052.1 | - | 26 | 3206 | |||
AY497053.1 | - | 22 | 2795 | |||
AY497054.1 | - | 21 | 2696 | |||
AY497055.1 | - | 24 | 2981 | |||
AY497056.1 | - | 21 | 2952 | |||
AY497057.1 | - | 20 | 2851 | |||
AY497058.1 | - | 20 | 2690 | |||
AY497059.1 | - | 22 | 3061 | |||
AY497060.1 | - | 18 | 2628 |