Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRWD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153252.4 | NP_694984.4 | 30 | 3703 | ||
XM_005262113.3 | XP_005262170.1 | 29 | 3555 | |||
XM_017029384.1 | XP_016884873.1 | 26 | 3233 | |||
GenBank mRNA | AK095887.1 | - | 13 | 1576 | ||
AY497046.1 | - | 30 | 3703 | |||
AY497047.1 | - | 27 | 3635 | |||
AY497048.1 | - | 30 | 3726 | |||
AY497049.1 | - | 29 | 3696 | |||
AY497050.1 | - | 30 | 3676 | |||
AY497051.1 | - | 27 | 3263 | |||
AY497052.1 | - | 32 | 3862 | |||
AY497053.1 | - | 28 | 3451 | |||
AY497054.1 | - | 27 | 3352 | |||
AY497055.1 | - | 30 | 3637 | |||
AY497056.1 | - | 27 | 3608 | |||
AY497057.1 | - | 26 | 3507 | |||
AY497058.1 | - | 26 | 3346 | |||
AY497059.1 | - | 28 | 3717 | |||
AY497060.1 | - | 24 | 3284 | |||
BC039508.2 | - | 2 | 176 | |||
CB269573.1 | - | 1 | 77 |