Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHBP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099409.1 | NP_001092879.1 | 1 | 355 | ||
XM_005273867.3 | XP_005273924.1 | 1 | 358 | |||
XM_005273868.3 | XP_005273925.1 | 1 | 357 | |||
XM_005273869.3 | XP_005273926.1 | 1 | 359 | |||
XM_005273870.3 | XP_005273927.1 | 1 | 358 | |||
XM_006718486.3 | XP_006718549.1 | 1 | 356 | |||
XM_006718487.3 | XP_006718550.1 | 1 | 356 | |||
XM_017017467.1 | XP_016872956.1 | 1 | 77 | |||
XM_017017469.1 | XP_016872958.1 | 1 | 357 | |||
GenBank mRNA | AL834433.1 | - | 1 | 52 | ||
BX443367.2 | - | 1 | 355 |