Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203301.3 | NP_976046.1 | 3 | 1533 | ||
GenBank mRNA | AK294140.1 | - | 1 | 377 | ||
BC042535.1 | - | 1 | 12 | |||
BC053537.1 | - | 3 | 1449 | 5181903 | ||
BC068566.1 | - | 3 | 1505 | 5270962 |