Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100876.1 | NP_001094346.1 | 4 | 1090 | ||
NM_001100877.1 | NP_001094347.1 | 2 | 198 | |||
NM_174933.3 | NP_777593.2 | 4 | 1090 | |||
GenBank mRNA | AK054994.1 | - | 5 | 602 | ||
AK095000.1 | - | 4 | 1090 | |||
BC009373.2 | - | 4 | 577 | 4121964 | ||
BC051300.1 | - | 2 | 170 | 4843267 | ||
BC053853.1 | - | 2 | 159 | 5209351 |