Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286828.1 | NP_001273757.1 | 3 | 377 | ||
NM_198469.3 | NP_940871.2 | 4 | 489 | |||
XM_005251877.4 | XP_005251934.1 | 4 | 485 | |||
XM_005251878.3 | XP_005251935.1 | 4 | 460 | |||
XM_006717034.3 | XP_006717097.1 | 3 | 425 | |||
XM_011518457.2 | XP_011516759.1 | 4 | 485 | |||
XM_011518458.1 | XP_011516760.1 | 5 | 568 | |||
GenBank mRNA | AK128877.1 | - | 4 | 486 | ||
AK302081.1 | - | 4 | 463 | |||
BC133690.1 | - | 4 | 480 | 40119769 | ||
BC143440.1 | - | 3 | 337 | |||
HY011325.1 | - | 4 | 490 |