Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206997.1 | NP_001193926.1 | 2 | 251 | ||
NM_152770.2 | NP_689983.2 | 2 | 251 | |||
XM_011531816.2 | XP_011530118.2 | 3 | 417 | |||
XM_017007972.1 | XP_016863461.1 | 3 | 403 | |||
XM_017007973.1 | XP_016863462.1 | 3 | 406 | |||
XM_017007974.1 | XP_016863463.1 | 3 | 411 | |||
XM_017007976.1 | XP_016863465.1 | 3 | 426 | |||
XM_017007977.1 | XP_016863466.1 | 3 | 417 | |||
GenBank mRNA | AK128642.1 | - | 2 | 203 | ||
BC034296.1 | - | 2 | 251 | 5165558 |