Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCOLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153259.3 | NP_694991.2 | 8 | 1201 | ||
XM_005270719.3 | XP_005270776.1 | 8 | 978 | |||
XM_006710552.2 | XP_006710615.1 | 9 | 1291 | |||
XM_011541187.2 | XP_011539489.1 | 8 | 1035 | |||
XM_011541188.2 | XP_011539490.1 | 8 | 1318 | |||
GenBank mRNA | AK129786.1 | - | 2 | 204 | ||
AK309858.1 | - | 8 | 1192 | |||
AY083533.1 | - | 7 | 867 | |||
BC104891.1 | - | 8 | 1019 | 8143894 | ||
BC104893.1 | - | 8 | 1019 | 8143896 |