Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153260.2 | NP_694992.2 | 4 | 562 | ||
XM_011521423.2 | XP_011519725.1 | 3 | 450 | |||
XM_011521424.2 | XP_011519726.1 | 3 | 450 | |||
GenBank mRNA | AK094131.1 | - | 4 | 556 | ||
AK094891.1 | - | 3 | 796 | |||
AK309861.1 | - | 3 | 1127 | |||
BC058935.1 | - | 4 | 518 | 5578024 | ||
BX538324.1 | - | 3 | 708 | |||
CR749223.1 | - | 4 | 531 |