Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270989.1 | NP_001257918.1 | 3 | 259 | ||
NM_001270990.1 | NP_001257919.1 | 3 | 259 | |||
NM_001270991.1 | NP_001257920.1 | 3 | 232 | |||
NM_001270992.1 | NP_001257921.1 | 3 | 259 | |||
NM_001270993.1 | NP_001257922.1 | 3 | 232 | |||
NR_073114.1 | 3 | 259 | ||||
NR_073117.1 | 2 | 169 | ||||
XM_005265666.4 | XP_005265723.1 | 3 | 1131 | |||
XM_011531817.2 | XP_011530119.1 | 3 | 1117 | |||
GenBank mRNA | AK289455.1 | - | 3 | 257 | ||
AY358920.1 | - | 3 | 215 | |||
BC127938.1 | - | 3 | 255 | 40132636 | ||
BC127939.1 | - | 3 | 255 | 40132642 | ||
CB150835.1 | - | 3 | 232 | |||
DQ235264.1 | - | 3 | 195 | |||
DQ235265.1 | - | 3 | 168 | |||
DQ235266.1 | - | 2 | 105 | |||
DQ235267.1 | - | 3 | 195 | |||
DQ235269.1 | - | 3 | 243 |