Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF718 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039127.5 | NP_001034216.2 | 4 | 646 | ||
NM_001289930.1 | NP_001276859.1 | 4 | 1269 | |||
NM_001289931.1 | NP_001276860.1 | 4 | 571 | |||
NR_110529.1 | 2 | 423 | ||||
XM_017007979.1 | XP_016863468.1 | 5 | 2847 | |||
XM_017007980.1 | XP_016863469.1 | 5 | 2429 | |||
XM_017007981.1 | XP_016863470.1 | 5 | 676 | |||
GenBank mRNA | AK074517.1 | - | 1 | 55 | ||
AK096662.1 | - | 3 | 1301 | |||
AL833758.1 | - | 2 | 379 | |||
BC047874.1 | - | 1 | 581 | |||
BC104026.1 | - | 4 | 1207 | 40025602 | ||
BC104027.2 | - | 4 | 584 | 40025603 | ||
BC104028.1 | - | 4 | 1207 | 40025604 | ||
BC104029.2 | - | 4 | 584 | 40025606 | ||
BC154406.1 | - | 4 | 584 | 40112082 |