Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038440.1 | 2 | 172 | |||
GenBank mRNA | AK091766.1 | - | 1 | 155 | ||
CB992854.1 | - | 2 | 199 |