Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H2BA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170610.2 | NP_733759.1 | 1 | 206 | ||
GenBank mRNA | AK312055.1 | - | 1 | 338 | ||
BC066240.1 | - | 1 | 206 | 7002052 | ||
BC066242.1 | - | 1 | 206 | 7002055 | ||
BC066243.1 | - | 1 | 205 |