Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR52B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004052.1 | NP_001004052.1 | 1 | 927 | ||
GenBank mRNA | BC136732.1 | - | 1 | 1009 | 9020722 | |
BC136733.1 | - | 1 | 1009 | 9020723 |