Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPNT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033047.2 | NP_001028219.1 | 2 | 301 | ||
NM_001184690.1 | NP_001171619.1 | 2 | 301 | |||
NM_001184691.1 | NP_001171620.1 | 2 | 301 | |||
NM_001184692.1 | NP_001171621.1 | 2 | 301 | |||
NM_001184693.1 | NP_001171622.1 | 2 | 301 | |||
XM_005262888.4 | XP_005262945.1 | 2 | 288 | |||
XM_011531820.2 | XP_011530122.1 | 2 | 291 | |||
XM_011531822.2 | XP_011530124.1 | 2 | 291 | |||
XM_011531823.2 | XP_011530125.1 | 2 | 291 | |||
XM_011531824.2 | XP_011530126.1 | 2 | 291 | |||
XM_011531825.2 | XP_011530127.1 | 2 | 291 | |||
XM_017007984.1 | XP_016863473.1 | 2 | 291 | |||
GenBank mRNA | AK290029.1 | - | 2 | 248 | ||
AK295772.1 | - | 2 | 301 | |||
AK302477.1 | - | 2 | 300 | |||
AK304279.1 | - | 2 | 300 | |||
AK304721.1 | - | 2 | 248 | |||
AL832465.1 | - | 2 | 215 | |||
AY358336.1 | - | 2 | 290 | |||
BC131587.1 | - | 2 | 148 |