Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181844.3 | NP_862827.1 | 4 | 578 | ||
GenBank mRNA | AB076580.1 | - | 4 | 558 | ||
AK122893.1 | - | 4 | 574 | |||
AK292878.1 | - | 4 | 561 | |||
AK293102.1 | - | 4 | 577 | |||
BC059404.1 | - | 4 | 579 | 30342006 |