Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNEP1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281789.1 | NP_001268718.1 | 2, 3 | 210 | ||
NM_153261.5 | NP_694993.2 | 3, 4 | 261 | |||
GenBank mRNA | AK095420.1 | - | 2, 3 | 183 | ||
AK301610.1 | - | 3, 4 | 226 | |||
AK307132.1 | - | 2, 3 | 257 | |||
BC036683.2 | - | 3, 4 | 190 | 5266084 | ||
CR933600.1 | - | 3, 4 | 206 | |||
FJ516381.1 | - | 2, 3 | 138 | |||
HY011229.1 | - | 2, 3 | 211 |