Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164443.1 | NP_001157915.1 | 20, 21 | 4369 | ||
XM_011543298.2 | XP_011541600.1 | 21, 22 | 4562 | |||
XM_011543299.1 | XP_011541601.1 | 21, 22 | 4540 | |||
XM_011543300.2 | XP_011541602.1 | 21, 22 | 4562 | |||
XM_011543301.2 | XP_011541603.1 | 20, 21 | 4391 | |||
XM_011543302.2 | XP_011541604.1 | 19, 20 | 4098 | |||
XM_017009317.1 | XP_016864806.1 | 21, 22 | 4457 | |||
XM_017009318.1 | XP_016864807.1 | 21, 22 | 4562 | |||
XM_017009319.1 | XP_016864808.1 | 21, 22 | 4271 | |||
XM_017009320.1 | XP_016864809.1 | 18, 19 | 4083 | |||
XM_017009321.1 | XP_016864810.1 | 14, 15 | 3147 |